Tisk…
Cartridge filters, ProcessGard® CR
Dodavatel: ENTEGRIS
CR0101006 CR2501006 CR9901006 CR99F1006 CR1001006 CR0101V06 CR0101T06 CR0102006 CR0103006 CR0171006 CR0172006 CR0173006 CR01F1006 CR01F1P06 CR01F2006 CR0301006 CR0302006 CR03F2006 CR03F1006 CR03F1P06 CR03M0203 CR0501006 CR05010E6 CR0501E06 CR0501T06 CR0502006 CR0502T06 CR0503006 CR0571006 CR05F1006 CR10010V6 CR1001T06 CR1002006 CR1071006 CR10F1006 CR10F2006 CR25010E6 CR2502006 CR2503006 CR25F2006 CR5001T06 CR5003006 CR7501006 CRA5010E6 CRK102006 CRA502T06 CRA5F1006 CRA5F1P06 CRA5F2006 CRK3F1006
ENTECR0101006EA 0 CZK
ENTECR0101006 ENTECR2501006 ENTECR9901006 ENTECR99F1006 ENTECR1001006 ENTECR0101V06 ENTECR0101T06 ENTECR0102006 ENTECR0103006 ENTECR0171006 ENTECR0172006 ENTECR0173006 ENTECR01F1006 ENTECR01F1P06 ENTECR01F2006 ENTECR0301006 ENTECR0302006 ENTECR03F2006 ENTECR03F1006 ENTECR03F1P06 ENTECR03M0203 ENTECR0501006 ENTECR05010E6 ENTECR0501E06 ENTECR0501T06 ENTECR0502006 ENTECR0502T06 ENTECR0503006 ENTECR0571006 ENTECR05F1006 ENTECR10010V6 ENTECR1001T06 ENTECR1002006 ENTECR1071006 ENTECR10F1006 ENTECR10F2006 ENTECR25010E6 ENTECR2502006 ENTECR2503006 ENTECR25F2006 ENTECR5001T06 ENTECR5003006 ENTECR7501006 ENTECRA5010E6 ENTECRK102006 ENTECRA502T06 ENTECRA5F1006 ENTECRA5F1P06 ENTECRA5F2006 ENTECRK3F1006
Cartridge filters, ProcessGard® CR
Filtry Patrony filtrační
These cartridge filters are widely used in chemical, electronic and semiconductor industries for liquid and gas prefiltration applications. They retain contaminants by means of a multi-stage, graded density design.
  • Depth filter design
  • Superior retention of colloids and particles ensuring low particle counts in bulk DI water and chemical production
  • Excellent chemical and heat resistance
  • Manufactured, tested and packaged in an ISO 9001 Quality Systems Standard facility
Membrane: PP; Supports, cage, core, sleeves and endcaps: PP
Maximum differentialMore Product Information