Tisk…
Forceps
Dodavatel: ENTEGRIS
C20-06F12
ENTEC20-06F12EA 0 CZK
ENTEC20-06F12
Forceps
Pinzety
Glass-filled PP, white. Straight forceps for substrate handling.
  • Extra heavy for much stronger grip at ends
  • Blunt tips
  • Packaged in protective cases