Tisk…

Point of use photochemical filters, Optimizer®

Dodavatel: ENTEGRIS
ABUG0LEM1 ABUG0SKM1 ABUV0LEM1 ABUV0SEM1 ABUZ0LEM1 ABUZ0LKM1 ABUZ0SKM1 AHUV0LEM1 AHUV0LKM1 AHUV0SEM1 AHUV0SKM1 AHUYMLEM1 AHUYMLKM1 AHUZ0LEM1 AHUZ0LKM1 AHUZ0SEM1 AHUZ0SKM1 AWAV0SKM1 AWAXMSKM1
514-0733EA 0 CZK
514-0733 ENTEABUG0SKM1 ENTEABUV0LEM1 ENTEABUV0SEM1 ENTEABUZ0LEM1 ENTEABUZ0LKM1 ENTEABUZ0SKM1 ENTEAHUV0LEM1 ENTEAHUV0LKM1 ENTEAHUV0SEM1 ENTEAHUV0SKM1 ENTEAHUYMLEM1 ENTEAHUYMLKM1 ENTEAHUZ0LEM1 ENTEAHUZ0LKM1 ENTEAHUZ0SEM1 ENTEAHUZ0SKM1 ENTEAWAV0SKM1 ENTEAWAXMSKM1
Point of use photochemical filters, Optimizer®
Filtry Patrony filtrační
Liquid photochemical filters designed for developer, DI water and solvent applications in POU photolithography processes.

  • Advanced filtration media: optimises filtration and particle retention capabilities
  • UPE membrane spontaneously wets and stays wet in solvents

Membrane: Patented surface-modified hydrophilic ultra-high molecular weight polyethylene (UPE); Supports, core and sleeve: HDPE; Housing: PP

Maximum operating temperature: 40 °C

Maximum inlet pressure: 3,9 bar at 25 °C

Maximum forward/reverse pressure: 2,8 bar at 25 °C
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR