Tisk…
Elmasonic EASY Ultrasonic Baths
Dodavatel: ELMA SCHMIDBAUER
1118881 1118883 1118885 1118887 1117697 1118890 1118892 1118894 1118896 1118898 1118900 1118902
ELMA1118881EA 9350 CZK
ELMA1118881 ELMA1118883 ELMA1118885 ELMA1118887 ELMA1117697 ELMA1118890 ELMA1118892 ELMA1118894 ELMA1118896 ELMA1118898 ELMA1118900 ELMA1118902
Elmasonic EASY Ultrasonic Baths
Lázně ultrazvukové
The Elmasonic EASY ultrasonic bath series includes 9 units of different sizes and is characterised by simple, user-friendly operation. With 100% ultrasonic power at a frequency of 37 kHz, the EASY solves cleaning tasks quickly and efficiently. Ideal for removing tenacious, mineral soiling and polishing pastes.
  • 37 kHz ultrasonic frequency (control range −2,5 / +5,5 kHz)
  • 2 ultrasonic modes: dynamic mode or eco mode
  • Electronic selector switches on wipeable membrane keypad and LED display of all operating states
  • Electronic temperature control adjustable in
More Product Information