Tisk…
Cole-Parmer® HT-200 Grounded Heating Tapes
Dodavatel: Cole-Parmer
36220-10 36220-12 36220-00 36220-04 36220-06 36220-08 36220-02 36220-15 36220-14
ELET36220-10EA 30570 CZK
ELET36220-10 ELET36220-12 ELET36220-00 ELET36220-04 ELET36220-06 ELET36220-08 ELET36220-02 ELET36220-15 ELET36220-14
Cole-Parmer® HT-200 Grounded Heating Tapes
Pásky
Flexible heating tapes are designed to fit metal pipes and glassware.
  • Maximum element temperature of 842 °F (450 °C)
  • Braided earth wire beneath the outer sheath
  • Approximately 25 mm wide
The flexible fiberglass heating tapes are ideal for heating columns, pipes, valves, and transfer lines, where applications need to be observed. The heating element and braided ground wire are wound on a fiberglass core and covered in a braided fiberglass outer sleeve. Tapes are not water-resistant; use in dry conditions only. Tapes are supplied with 3,5'' (9 cm)More Product Information