Tisk…
Cole-Parmer® HM-200 Large Volume Heating Mantles
Dodavatel: Cole-Parmer
36013-16 36013-18 36013-20 36013-22
ELET36013-16EA 179170 CZK
ELET36013-16 ELET36013-18 ELET36013-20 ELET36013-22
Cole-Parmer® HM-200 Large Volume Heating Mantles
Ohřívače Hnízda topná
Accommodate 60° funnels, pear-shaped flasks and round-bottom flasks.
  • Bottom opening allows use with 60° funnels of various diameters
  • Accept large capacity flasks -between 10 to 22 litres
  • Three separate electrical circuits allow controlled heating of top, middle and bottom of flasks independently
The 'V-shaped' design of the heating mantle accommodates both pear-shaped and round bottom flasks. Coiled heating element, suspended within a thermal insulating material, provides optimal heat transfer and support. The outer casing remains cool to the touchMore Product Information