Tisk…
JALAS® GRAN PREMIO 1518 ANTISLIP+ ESD Safety Sandals
Dodavatel: EJENDALS
1518-49 1518-48 1518-50
EJEN1518-49EA 3890 CZK
EJEN1518-49 EJEN1518-48 EJEN1518-50
JALAS® GRAN PREMIO 1518 ANTISLIP+ ESD Safety Sandals
Obuv
Sandal that breathes, with a spacious last. With all the necessary safety features for those who prioritize comfort and cool feet. The sandal is ventilated and has a spacious last. Hook and loop closure, nail protection in steel and toe cap in aluminium.
  • ProNose toe reinforcement
  • Wide fit
  • Heat-resistant outsole
  • Oil-resistant outsole
  • Anti-static properties
  • Ventilating insole
  • Double shock absorption zones
  • Conforms with IEC 61340-5-1 (ESD)
These sandals has highest level of protection, excellent grip, perfect fit, extra comfortable, excellent shockMore Product Information