Tisk…
Chlorfenvinphos (2-Chloro-1-(2,4 dichlorophenyl) vinyl diethyl phosphate, mixture of cis and trans isomers) 100 µg/ml in cyclohexane
Dodavatel: EHRENSTORFER
XA11290000CY
EHERXA11290000CYEA 1180 CZK
EHERXA11290000CY
Chlorfenvinphos (2-Chloro-1-(2,4 dichlorophenyl) vinyl diethyl phosphate, mixture of cis and trans isomers) 100 µg/ml in cyclohexane
Standardy Standardy organické
Objednat
Produktová specifikace
Popis Chlorfenvinphos (2-Chloro-1-(2,4 dichlorophenyl) vinyl diethyl phosphate, mixture of cis and trans isomers)
Environmentally Preferable
Čí. CAS 470-90-6
Vzorec C₁₂H₁₄Cl₃O₄P