Tisk…

Glycerol phosphate disodium salt hydrate mixture of isomers

Dodavatel: EHRENSTORFER

TRC-G601603-1G
EHERTRC-G601603-1GEA 0 CZK
EHERTRC-G601603-1G
Glycerol phosphate disodium salt hydrate mixture of isomers
Glycerol phosphate disodium salt hydrate
Vzorec: C₃H₇Na₂O₆P·H₂O
Storage Temperature: Freezer
MDL Number: MFCD00149082
CAS číslo: 55073-41-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR