Tisk…

methyl 4-hydroxybenzoát

Dodavatel: EHRENSTORFER
Varování

Synonyms: Methylparaben, 4-Hydroxybenzoic acid methyl ester, Methyl Parahydroxybenzoate, Methyl 4-hydroxybenzoate

C14229000
EHERC14229000EA 1490 CZK
EHERC14229000
methyl 4-hydroxybenzoát
methyl 4-hydroxybenzoát
Vzorec: HOC₆H₄CO₂CH₃
MW: 152.15
Bod varu: 270…280 °C (1013 hPa)
Bod tání: 125…128 °C
Hustota: 1.1208 g/cm³ (137 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002352
CAS číslo: 99-76-3
EINECS: 202-785-7
Merck Index: 13,06129
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=84321052052f4be2a53d8d7ebac7fabe6fc5a435&vid=ede765fad275b2515a8c57ba4499bff313a18983&prid=web&clid=SAPDAM

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR