Tisk…

Ethyl 4-hydroxybenzoate

Dodavatel: EHRENSTORFER

Synonyms: Ethyl-4-hydroxybenzoat, Nipagin A, Ethyl paraben, Ethyl p-hydroxybenzoate, Ethyl-p-hydroxybenzoat, Ethylparaben

C14228800
EHERC14228800EA 1490 CZK
EHERC14228800
Ethyl 4-hydroxybenzoate
Ethyl 4-hydroxybenzoate
Vzorec: C₉H₁₀O₃
MW: 166,18 g/mol
Bod varu: 297 °C (1013 hPa)
Bod tání: 116…118 °C
Hustota: 1,078 g/cm³ (131 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002353
CAS číslo: 120-47-8
EINECS: 204-399-4
Merck Index: 13,03869

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR