Tisk…

2-Ethylphenol

Dodavatel: EHRENSTORFER

Synonyms: 1-Ethyl-2-hydroxybenzene

C13350000
EHERC13350000EA 1120 CZK
EHERC13350000
2-Ethylphenol
2-Ethylphenol
Vzorec: C₈H₁₀O
MW: 122.17
Bod varu: 204.5
Bod tání: –18 °C
Hustota: 1.0371
Storage Temperature: Ambient
MDL Number: MFCD00002249
CAS číslo: 90-00-6
EINECS: 201-958-4
UN: 3145
ADR: 8 III
Merck Index: 13,03871

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR