Tisk…

3-Nitro-o-xylene

Dodavatel: EHRENSTORFER

Synonyms: 1,2-Dimethyl-3-nitrobenzene, 2,3-Dimethylnitrobenzene, 3-Nitro-o-xylene

C12727900
EHERC12727900EA 1560 CZK
EHERC12727900
3-Nitro-o-xylene
3-Nitro-o-xylene
Vzorec: C₈H₉NO₂
Bod varu: 245 °C (1013 hPa)
Hustota: 1,1402 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007162
CAS číslo: 83-41-0
EINECS: 201-474-3
UN: 1665
ADR: 6.1,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR