Tisk…

Dichlorvos-[dimethyl-²H₆] (2,2-Dichlorovinyl dimethyl phosphate-[dimethyl-²H₆], DDVP-[dimethyl-²H₆])

Dodavatel: EHRENSTORFER

C12530100
EHERC12530100EA 21200 CZK
EHERC12530100
Dichlorvos-[dimethyl-²H₆] (2,2-Dichlorovinyl dimethyl phosphate-[dimethyl-²H₆], DDVP-[dimethyl-²H₆])
Dichlorvos-[dimethyl-²H₆] (2,2-dichlorovinyl dimethyl phosphate-[dimethyl-²H₆], DDVP-[dimethyl-²H₆])
Vzorec: C₄HCl₂D₆O₄P
CAS číslo: 203645-53-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR