Tisk…

p-Xylene-[D₁₀]

Dodavatel: EHRENSTORFER
Nebezpečí

Synonyms: p-Xylene-[²H₁₀], 1,4-Dimethylbenzene-D10

C12370100
EHERC12370100EA 2020 CZK
EHERC12370100
p-Xylene-[D₁₀]
p-Xylene-[D₁₀]
Vzorec: C₆D₄(CD₃)₂
Bod varu: 135 °C (1013 hPa)
Bod tání: 13,3 °C
Hustota: 0,948 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00037699
CAS číslo: 41051-88-1
EINECS: 255-193-6
UN: 1307
ADR: 3,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR