Tisk…
Experimentation kits, principles of thin layer chromatography
Dodavatel: EDVOTEK
113
EDVO113EA 3570 CZK
EDVO113
Experimentation kits, principles of thin layer chromatography
Materiály výukové Chemický výukový materiál Aktivity pro výuku chromatografie
This experiment introduces chromatographic theory and methods of thin layer chromatography (TLC). A mixture of dyes are separated on a cellulose-based TLC plate using different solvent systems.
  • For 8 lab groups
  • Spotting plates 20 minutes, TLC separation 5 minutes
Also required: 5ml or 10 ml pipettes.
Informace o dodávce: Kit includes instructions, samples, reagents and solvents, cellulose TLC plate, 5 µl capillary pipettes.