Tisk…
Organic reference standards, fosinoprils, Ph. Eur.
Dodavatel: EDQM
Y0001212 Y0001227 Y0001228 Y0001229 Y0001230 Y0001231 Y0001232 Y0001233
EDQMY0001212EA 6990 CZK
EDQMY0001212 EDQMY0001227 EDQMY0001228 EDQMY0001229 EDQMY0001230 EDQMY0001231 EDQMY0001232 EDQMY0001233
Organic reference standards, fosinoprils, Ph. Eur.
Standardy Standardy organické
Objednat
Produktová specifikace
Popis Fosinopril sodium Fosinopril impurity A, (2S,4S)-4-cyclohexyl-1-[[(R)-hydroxy(4-phenylbutyl)phosphoryl]acetyl]pyrrolidine-2-carboxylic acid Fosinopril impurity B, (2RS,4RS)-4-cyclohexyl-1-[[(RS)-[(1SR)-2-methyl-1-(1-oxopropoxy)propoxy](4-phenylbutyl)phosphoryl]acetyl]pyrrolidine-2-carboxylic acid Fosinopril impurity D, (2S,4R)-4-cyclohexyl-1-[[(R)-[(1S)-2-methyl-1-(1-oxopropoxy)propoxy](4-phenylbutyl)phosphoryl]acetyl]pyrrolidine-2-carboxylic acid Fosinopril impurity I, [(RS)-[(1SR)-2-methyl-1-(1-oxopropoxy)propoxy](4-phenylbutyl)phosphoryl]acetic acid
Environmentally Preferable
Čí. CAS 88889-14-9 95399-71-6 1356932-15-4 1356446-78-0 123599-82-6
Vzorec C₃₀H₄₅NNaO₇P C₂₃H₃₄NO₅P C₃₀H₄₅NNaO₇P C₃₀H₄₆NO₇P C₁₉H₂₉O₆P