Tisk…
Cleanroom bouffant caps, Tyvek® IsoClean®, Model IC729B WS
Dodavatel: DuPont
IC729SWHWS
DUPOIC729SWHWSEA 14930 CZK
DUPOIC729SWHWS
Cleanroom bouffant caps, Tyvek® IsoClean®, Model IC729B WS
Jednorázové oděvy
These bouffant caps, model IC729SWH WS, are made of Tyvek®, which is a fabric composed of flash spun high density polyethylene, providing an ideal balance of protection, durability, and comfort. Tyvek® is permeable to both air and water vapour, yet repels water-based liquids and aerosols. It provides an inherent barrier to particles, microorganisms, and non-hazardous light liquid splash.
  • Ultra-low linting fabric
  • Serged seams
  • Elastic headband
  • Gamma irradiated to Sterility Assurance Level (SAL) of 10⁻⁶
  • Suitable for use in GMP class A/B (ISO Class 5)
More Product Information