Tisk…
Gas cylinder storage boxes
Dodavatel: DUPERTHAL
7.90.100
DUEH7.90.100EA 21940 CZK
DUEH7.90.100
Gas cylinder storage boxes
Nábytek kancelářský Skříně pro zásoby
Storage box is used for storing gas cylinders outdoors in accordance with the regulations.
  • Galvanised sheet steel
  • Cylinder lock as standard fitting
  • Disassembled upon delivery – easy to assemble
Certifikace: Conforms with TRGS 510.
Informace o dodávce: Supplied with intermediate shelf.