Tisk…
Respirator filters for X-plore® 8000
Dodavatel: Dräger
6739535 6739545 6739550 6739555 6739580 6739585
DRAE6739535EA 1020 CZK
DRAE6739535 DRAE6739545 DRAE6739550 DRAE6739555 DRAE6739580 DRAE6739585
Respirator filters for X-plore® 8000
Polomasky a filtrační respirátory Polomasky filtrační opakovatelně použitelné Filtry pro masky
The X-plore® 8000 filter series offers an extensive programme of various filter types for all essential application fields and protects against a number of gases and vapours.