Tisk…
Biocide, ClearKlens Bi-Spore
Dodavatel: Diversey
7516197 7515828 100827588
DIVY7516197EA 7810 CZK
DIVY7516197 DIVY7515828 DIVY100827588
Biocide, ClearKlens Bi-Spore
Prostředky čistící Dezinfekční prostředek
ClearKlens Bi-Spore is a rapid action stable two part chlorine dioxide generating system, which passes the European sporicidal test (CEN 13704) in 5 minutes, well within realistic cleanroom surface drying times, 5 to 8 minutes after surface contact.
  • Passes CEN 1276, CEN 1650, CEN 13697 and CEN 13704
  • 0,2 µm filtered, aseptically filled and double bagged
  • Supplied with certificate of analysis and sterility where applicable
  • Complimentary rotational partner to ClearKlens Tego® 2000 and ClearKlens Cleansinald
ClearKlens Bi-Spore possesses aMore Product Information