Tisk…
EduCare™ Extraction Cleaner
Dodavatel: Diversey
101109395
DIVY101109395EA 1220 CZK
DIVY101109395
EduCare™ Extraction Cleaner
Prostředky čistící Detergenty a povrchové čističe
EduCare™ extraction cleaner is a powerful multi-purpose cleaner compatible with advanced generation fibers when used as an extraction cleaner.
  • Low-foaming for effective use in high pressure carpet extractors
  • Excellent hard water tolerance and performance in hot or cold water
  • Won't harm or fade carpet color because optical brighteners are not used
  • Protects equipment with unique corrosion inhibitors
  • Reduces costs and storage space with economical, concentrated formula
  • Colourless with a floral fresh scent
  • Shelf life: Two years
- Specially formulated forMore Product Information