Tisk…
Disinfectant sprays, Des E, SoftCare™
Dodavatel: Diversey
101104048
DIVY101104048EA 5430 CZK
DIVY101104048
Disinfectant sprays, Des E, SoftCare™
Péče o pokožku Dezinfekce na ruce Kapalné a pěnové desinfekční prostředky na pleť
These SoftCare™ Des E disinfectant sprays, alcohol-based are effective against bacteria, virus and fungi. It is made up of 70% ethanol, which ensures instant hand hygiene.
  • Leave on product, so does not require water
  • Prevents infection
  • Frangrance free
  • Dye free
This disinfectant spray is ideal for use in healthcare and food processing areas.Liquid format suitable for spray application.
Upozornení: Use biocides safely. Always read the label and product information before use.