Tisk…
Grill cleaners, SURE®
Dodavatel: Diversey
100894371
DIVY100894371EA 2700 CZK
DIVY100894371
Grill cleaners, SURE®
Prostředky čistící Detergenty a povrchové čističe
Detergent suitable for daily cleaning of heavily soiled surfaces, such as ovens and grills.
  • Specially developed for daily cleaning
  • Viscous product has excellent adherence on vertical surfaces
  • Ease of rinse
  • No fragrance or dye
  • Contains plant-based ingredients from renewable sources
  • Decomposed by natural processes
  • No environmental hazard classifications
SURE® grill cleaner is part of a unique range of plant-based, Cleaning products that deliver the professional cleaning results you need - while being safe for people and kind to the environment. SURE®More Product Information