Tisk…
Optical DO Sensor Cable
Dodavatel: DISTEK
2700-0418
DISI2700-0418EA 0 CZK
DISI2700-0418
Optical DO Sensor Cable
Čerpadla Čerpadla peristaltická
Optical DO sensor cable that will work only with the optical DO sensors such as the Hamilton VisiFerm ECS optical DO sensor.