Tisk…
Spectral lamps, OSRAM
Dodavatel: DIDALAB
POD068380 POD068360 POD068400 POD068420 POD068440 POD068450 POD068480
DIDAPOD068380EA 23510 CZK
DIDAPOD068380 DIDAPOD068360 DIDAPOD068400 DIDAPOD068420 DIDAPOD068440 DIDAPOD068450 DIDAPOD068480
Spectral lamps, OSRAM
Osvětlení/Lampy Lampy spektrální
Nine pin socket spectral lamps with dimensions (H×Ø) 107×21 mm, operated with 1 Amp fixed current.