Tisk…
Urine container, with boric acid
Dodavatel: DELTALAB
409202.B
DELT409202.BEA 7820 CZK
DELT409202.B
Urine container, with boric acid
Systémy pro klinickou diagnostiku Clinical Sample Collection/Preparation Urine Sample Collection/Preparation
This container includes a boric acid preservative which allows the storage and transport of the urine sample to the laboratory.
  • The bacteria remain viable for 24 to 48 hours or more, depending on the type of microorganisms
  • Sterile container, sterilised by ethylene oxide
  • Boric acid quantity of 0,4 g
The boric acid supports the viability of the bacteria while preventing further multiplication. This preservative does not alter the characteristics of the urine and will not affect sample determination.
Upozornení: *Výrobky na této stránce nespadají doMore Product Information