Tisk…
Water sampling bottles, rectangular
Dodavatel: DELTALAB
282331 282330 282320 282321 282323 282323.BU 282324 282324.BU
DELT282331EA 1960 CZK
DELT282331 DELT282330 DELT282320 DELT282321 DELT282323 DELT282323.BU DELT282324 DELT282324.BU
Water sampling bottles, rectangular
Láhve Láhve na odběr vzorků vody
Polyethylene, translucent, with red screw cap.
  • Tamper evident red cap with an internal joint
  • Hermetical closure
  • Sterilised by radiation
  • Available with or without sodium thiosulfate
With liquid sodium thiosulphate (24 mg/L): ideal to analyse water meant for human consumption, swimming pools, and any other water where chlorine may modify the composition of the sample while being transported.
Without sodium thiosulphate: ideal to sample water for physico-chemical analysis, as well as for other sampling requiring sterile bottles.
The minor sides areMore Product Information