Tisk…
Drums with screw cap
Dodavatel: DELTALAB
202605 202610 202625
DELT202605EA 246 CZK
DELT202605 DELT202610 DELT202625
Drums with screw cap
Sudy, soudky, bubny
Drums, made of HDPE, provided with screw cap with grip. Can be used for the transport of hazardous, toxic and noxious products.
  • Stackable, with security cap
  • Good chemical resistance
  • Four flat sides for sticking information labels