Tisk…
Racks for microcentrifuge tubes
Dodavatel: DELTALAB
200420
DELT200420EA 2810 CZK
DELT200420
Racks for microcentrifuge tubes
Stojany Tube Racks
PP, Two level.
  • Interlocking - can be extended lengthways
  • Two levels for easy manipulation of the tubes
  • Easy to read numbers for better tube viewing and identification.