Tisk…
Buckets, Round
Dodavatel: DELTALAB
19907 19905 19906
DELT19907EA 767 CZK
DELT19907 DELT19905 DELT19906
Buckets, Round
Kbelíky
HDPE, white.
  • Good chemical and temperature resistance
  • Moulded-in graduations
  • With spout
For industrial and laboratory applications. With red stainless metal handle. Very sturdy construction.