Tisk…
Cleaning booster, deconex® 32 EMULGATOR
Dodavatel: Borer Chemie
502800.00-KK5W
DECO502800.00-KK5WEA 4220 CZK
DECO502800.00-KK5W
Cleaning booster, deconex® 32 EMULGATOR
Prostředky čistící Detergenty a povrchové čističe
Liquid cleaning booster with emulsifying effect. For use in laboratory washers.
  • Rinsable without residues and guarantees therefore consistent and error-free results
  • With a broad cleaning spectrum, as booster in combination with alkaline or acidic deconex® cleaner
  • Particularly well suited for emulsifying of organic residues such as greases and oils, recommended against stubborn deposits
  • Perfect results even at low concentrations
  • Applicable in the pre- and main-washing cycle
  • pH-neutral and environmentally friendly