Tisk…
Powered air purifying respirators (PAPR), Airvisor 2
Dodavatel: HONEYWELL SAFETY
1013939 1013980
DALL1013939EA 13000 CZK
DALL1013939 DALL1013980
Powered air purifying respirators (PAPR), Airvisor 2
Polomasky a filtrační respirátory Systémy polomasek
Airvisor 2 MV Chemical Kit with acetate chemical resistant visor, two disposable visor covers, fabric hood and neck cover, waist belt complete, carry bag. For applications exposed to chemicals.
Airvisor 2 MV Industry Kit with polycarbonate impact resistant visor, one fabric cassette, two disposable visor covers and head cover, waist belt complete, carrying bag. For industrial applications.
  • Lightweight product with wide field of vision
  • Hygienical single use head and neck protection
  • Visor keeps its original appearance due to visor cover
  • Carbon filter element