Tisk…
Erlenmeyer flasks, PETG, with vented cap, Corning®
Dodavatel: Corning
431280 431281 431282 431283 431284 431285
CORN431280EA 7610 CZK
CORN431280 CORN431281 CORN431282 CORN431283 CORN431284 CORN431285
Erlenmeyer flasks, PETG, with vented cap, Corning®
Baňky Baňky dle Erlenmeyera
PETG, optically clear, with vented cap.
  • Flasks will not break when dropped
  • Designed for single-use, as flasks will collapse if autoclaved
  • Molded-in graduations for accuracy
  • Individually packaged and radiation sterilised
  • Sterility Assurance Level (SAL) of 10⁻⁶
  • Nonpyrogenic
  • Baffled or plain bottom available in each size
These wide-mouth flasks are available in 2000, 3000 and 5000 ml sizes with either a baffled or plain bottom. The flasks are constructed of USP Class VI PETG for excellent optical clarity and mechanical strength. Molded graduations areMore Product Information