Tisk…
Pipette tips station, Corning® DeckWorks™
Dodavatel: Corning
4141 4142 4143 4144
CORN4141EA 10410 CZK
CORN4141 CORN4142 CORN4143 612-4196
Pipette tips station, Corning® DeckWorks™
Špičky pipetovací
These tip stations maximise bench space while minimising plastic waste. The stations contain 960 tips, available in extended length 10 µl and traditional 200 µl sizes.
  • Ideal single and multi-channel pipetting stations
  • Five-layer deck design (2×96) matches the microtiter plate format
  • Stable pipetting platform fits easily into sterile hood environments
  • Lids can be snap-positioned to expose a single side (96 tips) only
Applications include pre- and post-PCR procedures, cell culture and assays requiring large tip quantities. These tip stations are offeredMore Product Information