Tisk…
Cleanroom wipes, pre-saturated, SATWIPES® Theta
Dodavatel: CONTEC
SWC10004
CONTSWC10004EA 7760 CZK
CONTSWC10004
Cleanroom wipes, pre-saturated, SATWIPES® Theta
Ubousky/Utěrky Utěrky pro čisté prostory Ubrousky vlhčené do čistých prostor
Presaturated wipes can offer many benefits over bottled alcohol and dry wipes for cleanroom use. Contec’s sterile SATWIPES Theta with 70% denatured ethanol consist of a perforated roll of presaturated wipes which are dispensed through a convenient centre pull canister.
  • Made of 55% cellulose and 45% polyester creating a highly sorbent and durable wipe
  • Presaturated wipes eliminate the need for trigger spray bottles
  • Low levels of particles and fibres
  • Resealable canister prevents wipes drying out during use
  • Linear tear bags are easy to open even when wearing
More Product Information