Tisk…
Cleaning swabs, sealed, CONSTIX® SP-9
Dodavatel: CONTEC
SP-9
CONTSP-9EA 3310 CZK
CONTSP-9
Cleaning swabs, sealed, CONSTIX® SP-9
Tampóny odběrové Tampóny pro čištění
The SP-9 sealed polyester swab consists of 100% knitted polyester fabric that is thermally bonded to a flex-tip PP handle.
  • Durable and absorbent
  • Features extremely low particles and residue levels
  • Polyester head provides enhanced sorbency, particle entrapment and removal
  • Polyester head make the swab compatible with most common solvents, such as alcohols and ketones
  • PP handle has a thin, flexible tip that enables access to very small areas and components
The swab head is made of cleanroom laundered; double-layer knitted microfibre polyester material.More Product Information