Tisk…
Cleaning swabs, sealed foam, CONSTIX® SF-17
Dodavatel: CONTEC
SF-17
CONTSF-17EA 2940 CZK
CONTSF-17
Cleaning swabs, sealed foam, CONSTIX® SF-17
Tampóny odběrové Tampóny pro čištění
The SF-17 sealed foam swab consists of 100 PPI sealed closed-cell polyurethane foam head formed over the cotton bud of a 6" applicator with wooden handle.
  • Soft polyurethane foam head provides good particle entrapment and removal
  • Wooden handle is solvent resistant and stiff enough to allow vigorous swabbing
  • No adhesives are used
The absorbent foam enables solvents, such as isopropanol and ethanol, to be readily absorbed for applications involving solvent delivery and removal. SF-17 swab is well suited to applying solutions and adhesives and is low inMore Product Information