Tisk…
Utěrky pro čisté prostory, Amplitude™ Prozorb®
Dodavatel: CONTEC
PZ-99 PZ-1212
115-0272EA 10440 CZK
115-0272 CONTPZ-1212
Utěrky pro čisté prostory, Amplitude™ Prozorb®
Ubousky/Utěrky Utěrky pro čisté prostory Ubrousky nevlhčené do čistých prostor
Amplitude™ Prozorb® wipes are a hydroentangled blend of cellulose and polyester, produced through a special process that creates apertures in the fabric while enhancing the thickness and providing excellent cleanliness.
  • Durable textured surface promotes cleaning and particle removal capability
  • Excellent sorbency, great level of intrinsic sorbency and quick rate of sorbency
  • Low level of particles and extractables
Ideal for spill clean up in critical environments, biofilm and residue removal, product wipe down prior to cleanroom entry and wiping to dry.More Product Information