Tisk…
QuickConnect™ Mop Handles
Dodavatel: CONTEC
CFH32 CFH70 CFH114 CFH224 CFH334
CONTCFH32EA 3740 CZK
CONTCFH32 CONTCFH70 CONTCFH114 CONTCFH224 CONTCFH334
QuickConnect™ Mop Handles
Mopy
Interchangeable mop handles, available in carbon fiber, anodised aluminum, stainless steel, or fiberglass.
  • Compatible with all QuickConnect™ frames
  • Multiple lengths available
Fully autoclavable QuickConnect™ mop handles and interchangeable mop head frames work together for maximum flexibility when cleaning in critical environments. Invest in one handle and the various interchangeable heads required for specific applications. Handles are available in several lengths and styles.
Telescoping handles are available in lightweight carbon fiber, as wellMore Product Information