Tisk…
Roll mop, PolyTack®
Dodavatel: CONNECTICUT CLEANROOM
PH-18 PR-18F PR-9F PR-18
CONNPH-18EA 8190 CZK
CONNPH-18 CONNPR-18F CONNPR-9F CONNPR-18
Roll mop, PolyTack®
Mopy
Tacky roller, with foam or film for cleaning walls, floors, ceiling and hard to reach areas.
  • Easy to use hand or pole attachments
  • Plastic inner core insures cleanliness
  • Anti-microbial agent providing protection against bacterial growth