Tisk…
Acetic Acid (31)
Dodavatel: CUSTOM MADE CHEMICALS LAB
JPH123
CMPLJPH123EA 0 CZK
CMPLJPH123
Acetic Acid (31)
Standardy Standardy organické
The standard is specially intended for use in testing Japanese pharmacopoeia and monographs. This solution was produced according of Japanese pharmacopoeia specifications using reagent quality components and water meeting the Japanese pharmacopoeia monograph requirement for purified water.
Certifikace: ISO 17034, 17025, 9001 certified.