Tisk…

Dusičnan ceričito-amonný 0.1 mol/l (0.1 N) vodný roztok Reag. Ph. Eur.

Dodavatel: CUSTOM MADE CHEMICALS LAB
Varování

Synonyms: Ammonium ceric nitrate, Ammonium hexanitratocerate (IV), Ceric Ammonium Nitrate, Cerium (IV) ammonium nitrate, Diammonium hexanitratocerate

3000100
CMPL3000100EA 0 CZK
CMPL3000100
Dusičnan ceričito-amonný 0.1 mol/l (0.1 N) vodný roztok Reag. Ph. Eur.
Dusičnan ceričito-amonný
Vzorec: (NH₄)₂Ce(NO₃)₆
MW: 548.22
MDL Number: MFCD00151121
CAS číslo: 16774-21-3
EINECS: 240-827-6
Merck Index: 13,00509

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR