Tisk…
Pallets and sumps for storage of hazardous materials, GRP
Dodavatel: CEMO
7086 7717 5113 5114 7963 7970 7867 7868 7869 6882 6875 6886 7085 7083 7286 6884 6876 6943
CEMG7086EA 5310 CZK
CEMG7086 CEMG7717 CEMG5113 CEMG5114 CEMG7963 CEMG7970 CEMG7867 CEMG7868 CEMG7869 CEMG6882 CEMG6875 CEMG6886 CEMG7085 CEMG7083 CEMG7286 CEMG6884 CEMG6876 CEMG6943
Pallets and sumps for storage of hazardous materials, GRP
Skladování chemikálií
The sumps are applicable for the storage of environmentally hazardous substances.
The thermosetting plastic base structure in conjugation with the glass reinforcement provide an advantageous fire behaviour and high chemical resistance.