Tisk…
Steel ground protection systems
Dodavatel: CEMO
7401 7402 10617 7403 7404 7405 10618 7406
CEMG7401EA 27240 CZK
CEMG7401 CEMG7402 CEMG10617 CEMG7403 CEMG7404 CEMG7405 CEMG10618 CEMG7406
Steel ground protection systems
Absorpční materiály
These ground protection systems ensures safe handling of water-polluting as well as flammable substances - previously classed as AI*, AII, AIII and B.
  • Storage of water-polluting and flammable substances
  • Painted steel RAL 5002, ultramarine blue
  • Hot-dip galvanised gratings
Modular matched elements enable the lining of any large surfaces as a storage or transfilling station.