Tisk…

17-Octadecynoic acid

Dodavatel: Cayman Chemical

90270-1 90270-10 90270-100 90270-5
CAYM90270-1EA 675 CZK
CAYM90270-1 CAYM90270-10 CAYM90270-100 CAYM90270-5
17-Octadecynoic acid
17-Octadecynoic acid
Vzorec: C₁₈H₃₂O₂
CAS číslo: 34450-18-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR