Tisk…

4-Deoxypyridoxine hydrochloride ≥98%, crystalline solid

Dodavatel: Cayman Chemical
Varování

Synonyms: 5-(hydroxymethyl)-2,4-dimethylpyridin-3-ol hydrochloride, 5-Hydroxy-4,6-dimethyl-3-pyridinemethanol Hydrochloride, 4-Deoxypyridoxine HCl

21863-1 21863-100 21863-250 21863-500
CAYM21863-1EA 8090 CZK
CAYM21863-1 CAYM21863-100 CAYM21863-250 CAYM21863-500
4-Deoxypyridoxine hydrochloride ≥98%, crystalline solid
4-Deoxypyridoxine hydrochloride

4-Deoxypyridoxine (4-DPD) is a vitamin B6 antimetabolite with diverse biological activities. It inhibits transport of pyridoxine, pyridoxal, and pyridoxamine in and reduces growth of S. carlsbergensis cells. 4-DPD inhibits sphingosine-1-phosphate (S1P) lyase and reduces cyclic stretch-induced apoptosis in alveolar epithelial MLE-12 cells.

Vzorec: C₈H₁₁NO₂
MW: 189.64 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00051296
CAS číslo: 148-51-6

Objednat

Výsledky specifikačního testu

Purity ≥98%
λmax 286 nm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR