Tisk…

1-Methoxy-5-methylphenazinium methyl sulphate ≥98%, crystalline solid

Dodavatel: Cayman Chemical
Varování

21258-100 21258-25 21258-250 21258-50
CAYM21258-100EA 6990 CZK
CAYM21258-100 CAYM21258-25 CAYM21258-250 CAYM21258-50
1-Methoxy-5-methylphenazinium methyl sulphate ≥98%, crystalline solid
1-Methoxy-5-methylphenazinium methyl sulphate

1-Methoxy-5-methylphenazinium (methyl sulphate) is a photochemically stable mediator of electron transport between NADPH and various electron acceptors such as tetrazolium dyes. It can be used to visualise NAD-linked dehydrogenase activities.

Vzorec: C₁₅H₁₆N₂O₅S
Storage Temperature: Freezer
MDL Number: MFCD00040648
CAS číslo: 65162-13-2

Objednat

Výsledky specifikačního testu

Purity ≥98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR