Tisk…

N-Stearoyl-D-erythro-sphingosine 95, crystalline solid

Dodavatel: Cayman Chemical

Synonyms: C18 Ceramide (d18:1/18:0), (2S,3R,4E)-2-(Stearoylamino)-4-octadecene-1,3-diol, Cer(d18:1/18:0), N-[(1S,2R,3E)-2-Hydroxy-1-(hydroxymethyl)-3-heptadecen-1-yl]octadecanamide, C18-Ceramide, N-Stearoyl-D-sphingosine, N-Octadecanoyl-D-sphingosine

19556-10 19556-25 19556-5
CAYM19556-10EA 5610 CZK
CAYM19556-10 CAYM19556-25 CAYM19556-5
N-Stearoyl-D-erythro-sphingosine 95, crystalline solid
N-Stearoyl-D-erythro-sphingosine

C18 Ceramide is an endogenous bioactive sphingolipid. It is the primary short-chain ceramide found in brain tissue whose synthesis is regulated by longevity-assurance homologue 1 (Lass1) in mice.

Vzorec: C₃₆H₇₁NO₃
MW: 565.968 g/mol
Storage Temperature: Freezer
MDL Number: MFCD02259139
CAS číslo: 2304-81-6

Objednat

Výsledky specifikačního testu

Purity ≥95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR