Tisk…

FeTPPS ≥95%

Dodavatel: Cayman Chemical

17187-100 17187-250 17187-50
CAYM17187-100EA 3740 CZK
CAYM17187-100 CAYM17187-250 CAYM17187-50
FeTPPS ≥95%
FeTPPS

FeTPPS (Fe(III)5,10,15,20-tetrakis(4-sulfonatophenyl)porphyrinato chloride) is a ferric porphyrin complex that causes the decomposition of peroxynitrite by catalytic isomerization to produce nitrate both in vitro and in vivo.


  • Fe(III)5,10,15,20-tetrakis(4-sulfonatophenyl)porphyrinato chloride


The conversion of this reactive nitrogen species to nitrate results in cytoprotection (EC50 = 5 µM). FeTPPS does not complex with NO and does not alter superoxide directly. It is commonly used to elucidate the roles of peroxynitrite in oxidative stress, cell damage, and intracellular signaling.


Peroxynitrite is a highly reactive nitrogen species formed from the reaction of nitric oxide (NO) and superoxide.

Vzorec: C₄₄H₂₈ClFeN₄O₁₂S₄
MDL Number: MFCD00216780
CAS číslo: 90384-82-0

Objednat

Výsledky specifikačního testu

Purity ≥95%
Formulation A Crystalline Solid
UV/Vis λmax: 204, 324, 396, 530 nm
Stability ≥2 years at −20 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR